Quinaldic acid ≥98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 93-10-7 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: 156-159° Beilstein Reference: 126322 Merck Reference: 14,8046 MDL No.: MFCD00006752
Synonyms: Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
- CAS Number:93-10-7
- Formula:C₁₀H₇NO₂
- MDL Number:MFCD00006752
- Molecular Weight:173.17 g/mol
- Merck Index:14,8046
- Melting Pt:156…159 °C
- Storage Temperature:Ambient
- EINECS:202-218-3
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



