Order Entry
Export
Orders LinkContactUsLinkComponent
Quinaldic acid ≥98%

Quinaldic acid ≥98%

Glass bottle for solids, 5 g
Catalog # A10192.06
Irritant
Quinaldic acid ≥98%
Catalog # A10192.06
CAS Number:  93-10-7
Quinaldic acid ≥98%
Quinaldic acid ≥98%

Glass bottle for solids, 5 g

CAS Number:  93-10-7
Price
52,60 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 93-10-7 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: 156-159° Beilstein Reference: 126322 Merck Reference: 14,8046 MDL No.: MFCD00006752

Synonyms:  Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
  • CAS Number:93-10-7
  • Formula:C₁₀H₇NO₂
  • MDL Number:MFCD00006752
  • Molecular Weight:173.17 g/mol
  • Merck Index:14,8046
  • Melting Pt:156…159 °C
  • Storage Temperature:Ambient
  • EINECS:202-218-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications