Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dichlorobenzaldehyde ≥98%

2,3-Dichlorobenzaldehyde ≥98%

Glass bottle for solids, 250 g
Catalog # A10118.30
Corrosive
2,3-Dichlorobenzaldehyde ≥98%
Catalog # A10118.30
CAS Number:  6334-18-5
2,3-Dichlorobenzaldehyde ≥98%
2,3-Dichlorobenzaldehyde ≥98%

Glass bottle for solids, 250 g

CAS Number:  6334-18-5
Price
307,00 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:6334-18-5
  • Formula:C₇H₄Cl₂O
  • MDL Number:MFCD00010127
  • Molecular Weight:175.01 g/mol
  • Boiling Pt:143…145 °C (30 mmHg)
  • Melting Pt:61…65 °C
  • Density:1.33
  • Flash Pt:135 °C (275 °F)
  • Storage Temperature:Ambient
  • EINECS:228-711-3
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications