Order Entry
Export
Orders LinkContactUsLinkComponent
1,5-Pentanedithiol 96%

1,5-Pentanedithiol 96%

Glass bottle for solids, 5 g
Catalog # B25665.06
Irritant
1,5-Pentanedithiol 96%
Catalog # B25665.06
CAS Number:  928-98-3
1,5-Pentanedithiol 96%
1,5-Pentanedithiol 96%

Glass bottle for solids, 5 g

CAS Number:  928-98-3
Price
27,70 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 96% CAS No.: 928-98-3 Molecular Formula: HS(CH2)5SH Formula Weight: 136.28 Flash Point: 95°(203°F) Refractive Index: 1.5194 Odour: Unpleasant Melting Point: -72° Boiling Point: 216-218° Beilstein Reference: 1732335 UN No.: 3334 MDL No.: MFCD00004908

Synonyms:  1,5-Dimercaptopentane
  • CAS Number:928-98-3
  • Formula:C₅H₁₂S₂
  • MDL Number:MFCD00004908
  • Molecular Weight:136.28 g/mol
  • Boiling Pt:216…218 °C
  • Melting Pt:–72 °C
  • Density:1.016
  • Flash Pt:95 °C (203 °F)
  • Storage Temperature:Ambient
  • EINECS:213-194-9
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications