Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium peroxide 95%

Sodium peroxide 95%

Glass bottle for solids, 1.000 g
Catalog # L13306.0B
Oxidizing
Corrosive
Sodium peroxide 95%
Catalog # L13306.0B
CAS Number:  1313-60-6
Sodium peroxide 95%
Sodium peroxide 95%

Glass bottle for solids, 1.000 g

CAS Number:  1313-60-6
Price
612,00 €
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 95% CAS No.: 1313-60-6 Molecular Formula: Na2O2 Formula Weight: 77.98 Melting Point: 460° dec. Merck Reference: 14,8655 MDL No.: MFCD00003497

Synonyms:  di-Sodium peroxide
  • CAS Number:1313-60-6
  • Formula:Na₂O₂
  • MDL Number:MFCD00003497
  • Molecular Weight:77.98 g/mol
  • Merck Index:14,8655
  • Melting Pt:460 °C (dec.)
  • Density:2.805
  • Storage Temperature:Ambient
  • EINECS:215-209-4
  • UN:1504
  • ADR:5.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications