Order Entry
Export
Orders LinkContactUsLinkComponent
Methanol ≥99%

Methanol ≥99%

Glass bottle, 2.500 mL
Catalog # L13255.0F
Flammable
Toxic
Health
Methanol ≥99%
Catalog # L13255.0F
CAS Number:  67-56-1
Methanol ≥99%
Methanol ≥99%

Glass bottle, 2.500 mL

CAS Number:  67-56-1
Price
48,20 €
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04 Flash Point: 11°(52°F) Refractive Index: 1.3290 Odour: Mild Melting Point: -98° Boiling Point: 64.7° Beilstein Reference: 1098229 Merck Reference: 14,5957 UN No.: 1230 MDL No.: MFCD00004595

Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:14,5957
  • Boiling Pt:64.7 °C
  • Melting Pt:–98 °C
  • Density:0.791
  • Flash Pt:11 °C (52 °F)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications