Order Entry
Export
Orders LinkContactUsLinkComponent
Dimethyldichlorosilane 99%, rods

Dimethyldichlorosilane 99%, rods

Glass bottle for solids, 1.000 g
Catalog # L12133.0B
Flammable
Toxic
Corrosive
Irritant
Dimethyldichlorosilane 99%, rods
Catalog # L12133.0B
CAS Number:  75-78-5
Dimethyldichlorosilane 99%, rods

Glass bottle for solids, 1.000 g

CAS Number:  75-78-5
Price
119,00 €
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 75-78-5 Molecular Formula: C2H6Cl2Si Formula Weight: 129.06 Flash Point: -5°(23°F) Refractive Index: 1.4045 Melting Point: ca -76° Boiling Point: 70-71° Beilstein Reference: 605287 UN No.: 1162 MDL No.: MFCD00000491

Synonyms:  Dichloro(dimethyl)silane, Dichlorodimethylsilane
  • CAS Number:75-78-5
  • Formula:C₂H₆Cl₂Si
  • MDL Number:MFCD00000491
  • Molecular Weight:129.06 g/mol
  • Boiling Pt:70…71 °C
  • Melting Pt:∼–76 °C
  • Density:1.068
  • Flash Pt:–5 °C (23 °F)
  • Storage Temperature:Refrigerator
  • EINECS:200-901-0
  • UN:1162
  • ADR:3,II
  • REACH:01-2119437250-51

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications