Order Entry
Export
Orders LinkContactUsLinkComponent
Toluene ≥99%

Toluene ≥99%

Glass bottle, 2.500 mL
Catalog # L10967.0F
Flammable
Health
Irritant
Toluene ≥99%
Catalog # L10967.0F
CAS Number:  108-88-3
Toluene ≥99%
Toluene ≥99%

Glass bottle, 2.500 mL

CAS Number:  108-88-3
Price
62,90 €
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 108-88-3 Molecular Formula: C7H8 Formula Weight: 92.14 Flash Point: 4°(40°F) Refractive Index: 1.4967 Odour: Aromatic Melting Point: ca -93° Boiling Point: 111° Beilstein Reference: 635760 Merck Reference: 14,9529 UN No.: 1294 MDL No.: MFCD00008512

Synonyms:  Methylbenzene, Phenylmethane, FM approval
  • CAS Number:108-88-3
  • Formula:C₆H₅CH₃
  • MDL Number:MFCD00008512
  • Molecular Weight:92.14 g/mol
  • Merck Index:14,9529
  • Boiling Pt:111 °C
  • Melting Pt:∼–93 °C
  • Density:0.867
  • Flash Pt:4 °C (40 °F)
  • Storage Temperature:Ambient
  • EINECS:203-625-9
  • UN:1294
  • ADR:3,II
  • REACH:05-2114615130-69
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications