Orders LinkContactUsLinkComponent
(±)-α-Lipoic acid ≥98%
Glass bottle for solids, 25 g
  L04711.14
(±)-α-Lipoic acid ≥98%
  L04711.14
 :  1077-28-7
(±)-α-Lipoic acid ≥98%
(±)-α-Lipoic acid ≥98%
Glass bottle for solids, 25 g
 :  1077-28-7
249,00 €
1 * 25 g
Purity: 98% CAS No.: 1077-28-7 Molecular Formula: C8H14O2S2 Formula Weight: 206.33 Storage Temperature: 2°C to 8°C Melting Point: 59-63° Boiling Point: 160-165° Beilstein Reference: 81853 Merck Reference: 14,9326 MDL No.: MFCD00005474
 :  5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
  •  :1077-28-7
  •  :C₈H₁₄O₂S₂
  •  :MFCD00005474
  •  :206.33 g/mol
  •  :14,9326
  •  :160…165 °C
  •  :59…63 °C
  •  :214-071-2
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g