Cyclopentane for HPLC
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 287-92-3 Molecular Formula: C5H10 Formula Weight: 70.14 Flash Point: -37°(-34°F) Physical Form: Liquid Density: 0.748 Refractive Index: 1.4060 Appearance: Colourless Odour: Petrol-like Melting Point: -95° Boiling Point: 47-49° Beilstein Reference: 1900195 Merck Reference: 14,2741 UN No.: 1146 MDL No.: MFCD00001356
For HPLC and Spectrophotometry
Synonyms: Pentamethylene
- CAS Number:287-92-3
- Formula:C₅H₁₀
- MDL Number:MFCD00001356
- Molecular Weight:70.13 g/mol
- Merck Index:14,2741
- Boiling Pt:47…49 °C
- Melting Pt:–95 °C
- Density:0.748
- Flash Pt:–37 °C (–34 °F)
- Storage Temperature:Ambient
- EINECS:206-016-6
- UN:1146
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



