Order Entry
Export
Orders LinkContactUsLinkComponent
Soda lime, with indicator

Soda lime, with indicator

Glass bottle for solids, 2,5 kg
Catalog # 44786.A4
Corrosive
Soda lime, with indicator
Catalog # 44786.A4
CAS Number:  8006-28-8
Soda lime, with indicator
Soda lime, with indicator

Glass bottle for solids, 2,5 kg

CAS Number:  8006-28-8
Price
798,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

CAS No.: 8006-28-8 Physical Form: 8-12 Mesh Sensitivity: Air Sensitive and Hygroscopic Odour: Odourless Merck Reference: 14,8569 UN No.: 1907 MDL No.: MFCD00134094

  • CAS Number:8006-28-8
  • MDL Number:MFCD00134094
  • Merck Index:14,8569
  • Storage Temperature:Ambient
  • UN:1907
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications