n-Hexadecane ≥99.5%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99.5+% CAS No.: 544-76-3 Molecular Formula: CH3(CH2)14CH3 Formula Weight: 226.45 Flash Point: 135°(275°F) Physical Form: Liquid Density: 0.773 Refractive Index: 1.4345 Appearance: Colourless Melting Point: 18° Boiling Point: 287° Beilstein Reference: 1736592 MDL No.: MFCD00008998
Synonyms: Hexadecane, Cetane
- CAS Number:544-76-3
- Formula:H₃C(CH₂)₁₄CH₃
- MDL Number:MFCD00008998
- Molecular Weight:226.45 g/mol
- Boiling Pt:287 °C
- Melting Pt:18 °C
- Density:0.773
- Flash Pt:135 °C (275 °F)
- Storage Temperature:Ambient
- EINECS:208-878-9
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



