Order Entry
Export
Orders LinkContactUsLinkComponent
Multi-element plasma standard solution, non metals, Specpure®

Multi-element plasma standard solution, non metals, Specpure®

Multi-element plasma standard solution, non metals, Specpure®, As, B, P, S, Se, Si, Te @ 100µg/ml, 5% HCl/tr HF, 500 mL
Catalog # 44267.AP
Multi-element plasma standard solution, non metals, Specpure®
Catalog # 44267.AP
CAS Number:  
Multi-element plasma standard solution, non metals, Specpure®
Multi-element plasma standard solution, non metals, Specpure®

Multi-element plasma standard solution, non metals, Specpure®, As, B, P, S, Se, Si, Te @ 100µg/ml, 5% HCl/tr HF, 500 mL

CAS Number:  
Price
498,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

As, B, P, S, Se, Si, Te at concentration 100 µg/ml in 5% HCL with a trace of HF.

Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00144997

Specifications

Specifications

  • Matrix:
    5% HCl/tr HF
  • Pk:
    500 mL
  • Description:
    Multi-element plasma standard solution, non metals, Specpure®, As, B, P, S, Se, Si, Te @ 100µg/ml
  • Cat. No.:
    44267.AP
  • MDL No.:
    MFCD00144997

Specifications