Order Entry
Export
Orders LinkContactUsLinkComponent
Oleic acid ≥99%

Oleic acid ≥99%

Glass bottle for solids, 100 g
Catalog # 31997.22
Irritant
Oleic acid ≥99%
Catalog # 31997.22
CAS Number:  112-80-1
Oleic acid ≥99%
Oleic acid ≥99%

Glass bottle for solids, 100 g

CAS Number:  112-80-1
Price
941,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 112-80-1 Molecular Formula: C18H34O2 Formula Weight: 282.47 Physical Form: Liquid Density: 0.887 Refractive Index: 1.4610 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Melting Point: 13-14° Boiling Point: 286°/100mm Beilstein Reference: 1726542 Merck Reference: 14,6828 MDL No.: MFCD00064242

Synonyms:  (9Z)-9-Octadecenoic acid
  • CAS Number:112-80-1
  • Formula:C₁₈H₃₄O₂
  • MDL Number:MFCD00064242
  • Molecular Weight:282.47 g/mol
  • Merck Index:14,6828
  • Boiling Pt:286 °C (100 mmHg)
  • Melting Pt:13…14 °C
  • Density:0.887
  • Flash Pt:180 °C
  • Storage Temperature:Ambient
  • EINECS:204-007-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications