Order Entry
Export
Orders LinkContactUsLinkComponent
4-Ethylpyrocatechol ≥98%

4-Ethylpyrocatechol ≥98%

Glass bottle for solids, 250 mg
  A12048.MD
Irritant
4-Ethylpyrocatechol ≥98%
  A12048.MD
 :  1124-39-6
4-Ethylpyrocatechol ≥98%
4-Ethylpyrocatechol ≥98%

Glass bottle for solids, 250 mg

 :  1124-39-6
 
40,00 €
Log in for availability
1 * 250 mg
 

Purity: 98% CAS No.: 1124-39-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 37-40° Boiling Point: 105-107°/0.5mm MDL No.: MFCD00015847

Synonyms:  4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
  • CAS Number:1124-39-6
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00015847
  • Molecular Weight:138.17 g/mol
  • Boiling Pt:105…107 °C (0.5 mmHg)
  • Melting Pt:37…40 °C
  • Storage Temperature:Ambient
  • EINECS:214-397-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg

Specifications