4-Ethylpyrocatechol ≥98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
4-Ethylpyrocatechol ≥98%
Catalog # A12048.06
Supplier: Thermo Fisher Scientific
CAS Number: 1124-39-6
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 1124-39-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 37-40° Boiling Point: 105-107°/0.5mm MDL No.: MFCD00015847
Synonyms: 4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
- CAS Number:1124-39-6
- Formula:C₈H₁₀O₂
- MDL Number:MFCD00015847
- Molecular Weight:138.17 g/mol
- Boiling Pt:105…107 °C (0.5 mmHg)
- Melting Pt:37…40 °C
- Storage Temperature:Ambient
- EINECS:214-397-5
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



