Orders LinkContactUsLinkComponent
Disposable cannulas
Disposable cannulas, G14, Green, 70 mm, +, 2×70 mm
Catalog # EHRS15886470
Supplier:   EHRHARDT SOEHNE
Disposable cannulas
Catalog # EHRS15886470
Supplier:  EHRHARDT SOEHNE
Disposable cannulas
Disposable cannulas
Disposable cannulas, G14, Green, 70 mm, +, 2×70 mm
Price
130,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Disposable cannulas.
  • Pyrogen-free, not toxic
  • With lancet cut
  • EO-sterilisation
Documents
Specifications
  • Ø×L:
    2×70 mm
  • Gauge:
    G14
  • Sterile:
    +
  • Colour:
    Green
  • Description:
    Disposable cannulas
  • Length:
    70 mm
  • Seq:
    11