Orders LinkContactUsLinkComponent
KPG stirrer seals
KPG stirrer seals
Stirrer seals, KPG, for micro stirrer, NS 14/23, for 6 mm shaft
Catalog # QVFLLSG09033A01
Supplier:   QVF LABORTECHNIK
KPG stirrer seals
Catalog # QVFLLSG09033A01
Supplier:  QVF LABORTECHNIK
KPG stirrer seals
Stirrer seals, KPG, for micro stirrer, NS 14/23, for 6 mm shaft
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Stirrer seals, KPG, for micro stirrer, NS 14/23, for 6 mm shaft
  • Seq:
    01