Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Chromatography, Resin, Sorbent, BioSepra, Q Ceramic HyperD 20 um, 20040-028, Ion Exchange 1 * 500 ml
Catalog # 555-0018
Supplier:
Cytiva (Formerly Pall Lab)
Expand image
Chromatography, Resin, Sorbent, BioSepra, Q Ceramic HyperD 20 um, 20040-028, Ion Exchange 1 * 500 ml
Catalog # 555-0018
Supplier:
Cytiva (Formerly Pall Lab)