Orders LinkContactUsLinkComponent
Slide mailers
Slide mailers
Slide mailers, White, PP, 2/4
Catalog # SARS86.972.150
Supplier:   SARSTEDT
Slide mailers
Catalog # SARS86.972.150
Supplier:  SARSTEDT
Slide mailers
Slide mailers
Slide mailers, White, PP, 2/4
Price
116,00 €
Log in for availability
1 * 500 items
Product Details & Documents
These mailing vessels are manufactured from polypropylene and is suitable for the safe transportation or storage of two glass microscope slides (or four if placed against each other).
  • The lid features a ventilation hole
Ordering information: Containers available with or without ventilation hole.
Packaging: 50 pieces per sachet.
Specifications
  • No. of slides:
    2/4
  • Colour:
    White
  • Description:
    Slide mailers
  • Material:
    PP
  • Seq:
    01