Slide mailers
Slide mailers with ventilation, White, PP, 2/4
SARS86.972
: SARSTEDT
These mailing vessels are manufactured from polypropylene and is suitable for the safe transportation or storage of two glass microscope slides (or four if placed against each other).
- The lid features a ventilation hole
Ordering information: Containers available with or without ventilation hole.
Packaging: 50 pieces per sachet.
- No. of slides:2/4
- Colour:White
- Description:Slide mailers with ventilation
- Material:PP
- Seq:02