Inoculating loops and needles
Innoculating loops, 10 µl, Blue, 48/peel pack
Catalog # SARS86.1562.050
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PS, gamma sterilised. Time-saving, safe and easy to use. For laboratories with high workloads.
Specifications
- Packed:48/peel pack
- Capacity:10 µl
- Colour:Blue
- Description:Innoculating loops