96-Well micro test plates
Micro test plate with V bottom, with lid, sterile, Clear, 96, +, 1/bag
Catalog # SARS82.1583.001
Supplier: SARSTEDT
Jump to:
Product Details & Documents
Clear PS
- Compatible with all common microplate dispensers, readers, and washers
- Excellent and consistent optical quality
- Optimal surface
- Uniform design
- Choice of flat, round or conical wells
Specifications
- No. of wells:96
- Packed:1/bag
- Sterile:+
- Colour:Clear
- Description:Micro test plate with V bottom, with lid, sterile
Accessories
Product Information
- Catalog #ColourAvailabilityPrice
- Lids for micro test plates, PS