Orders LinkContactUsLinkComponent
96-Well micro test plates
Micro test plate with U bottom, with lid, sterile, Clear, 96, +, 1/bag
  SARS82.1582.001
 :   SARSTEDT
96-Well micro test plates
  SARS82.1582.001
 :  SARSTEDT
96-Well micro test plates
Micro test plate with U bottom, with lid, sterile, Clear, 96, +, 1/bag
89,80 €
1 * 50 items
Clear PS
  • Compatible with all common microplate dispensers, readers, and washers
  • Excellent and consistent optical quality
  • Optimal surface
  • Uniform design
  • Choice of flat, round or conical wells
  • No. of wells:
    96
  • Packed:
    1/bag
  • Sterile:
    +
  • Colour:
    Clear
  • Description:
    Micro test plate with U bottom, with lid, sterile
Accessories
  •  
    Description
    Colour
  • SARS82.1584
    Lids for micro test plates, PS
    Clear
    1 * 100 items - 43,60 €