96-Well micro test plates
Micro test plate with U bottom, without lid, Clear, 96, 25/bag
Catalog # SARS82.1582
Supplier: SARSTEDT
Jump to:
Product Details & Documents
Clear PS
- Compatible with all common microplate dispensers, readers, and washers
- Excellent and consistent optical quality
- Optimal surface
- Uniform design
- Choice of flat, round or conical wells
Specifications
- No. of wells:96
- Packed:25/bag
- Colour:Clear
- Description:Micro test plate with U bottom, without lid
Accessories
Product Information
- Catalog #ColourAvailabilityPrice
- Lids for micro test plates, PS