Order Entry
Export
Orders LinkContactUsLinkComponent
Mailing container, with screw cap, square

Mailing container, with screw cap, square

Mailing container with absorbent liner, square, 117×35 mm, Ø opening: 28 mm, with cap, Natural
Catalog # SARS78.573
Supplier:   SARSTEDT
Mailing container, with screw cap, square
Catalog # SARS78.573
Supplier:  SARSTEDT
Mailing container, with screw cap, square

Mailing container with absorbent liner, square, 117×35 mm, Ø opening: 28 mm, with cap, Natural

Price
123,00 €
Log in for availability
1 * 250 items
Product Details & Documents

Containers for safe postage and transportation of clinical samples (blood, urine, swabs and faeces tubes).

  • Tight screw caps ensures the safety
  • Made of neutral HDPE
  • One side of each container can be printed
  • Available with or without absorbent liners

Suitable for 30 ml liquid sample and faeces tubes of 107ר25 mm.

Specifications
  • Colour:
    Natural
  • Description:
    Mailing container with absorbent liner, square, 117×35 mm, Ø opening: 28 mm, with cap
  • Seq:
    02