Microtubes with thread in special sterile packaging
Microcentrifuge tubes with skirted base, without knurls, sterile, 2,0 ml, 500/double bag, 2500/case, 2,0
Catalog # SARS72.664.711
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, translucent
- With or without skirted base
Each bag is sub-packaged into a second additional bag. Removing the outer bag leaves a complete and sterile bag packaging for convenient processing in clean rooms or sterile environments. This packaging option is ideal for users working under clean room conditions to fill precious reagents into tubes.
Specifications
- Packed:500/double bag, 2500/case
- Recommended working volume:2,0
- Capacity:2,0 ml
- Description:Microcentrifuge tubes with skirted base, without knurls, sterile
Accessories
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.720Micro tubesYellowLog in for availability1 * 5.000 items - 341,00 €
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.721Micro tubesRedLog in for availability1 * 5.000 items - 341,00 €
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.723Micro tubesBlueLog in for availability1 * 5.000 items - 341,00 €
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.725Micro tubesNeutralLog in for availability1 * 5.000 items - 358,00 €
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.726Micro tubesGreenLog in for availability1 * 5.000 items - 341,00 €
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.727Micro tubesOrangeLog in for availability1 * 5.000 items - 288,00 €
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.729Micro tubesBrownLog in for availability1 * 5.000 items - 288,00 €
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.728Micro tubesBlackLog in for availability1 * 5.000 items - 288,00 €
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.757Micro tubesWhiteLog in for availability1 * 5.000 items - 288,00 €
Product Information
- Catalog #ForColourAvailabilityPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.755Micro tubesVioletLog in for availability1 * 5.000 items - 288,00 €