Orders LinkContactUsLinkComponent
Microtubes with thread in special sterile packaging
Microcentrifuge tubes with skirted base, sterile, 2,0 ml, 500/double bag, 2500/case
Catalog # SARS72.609.711
Supplier:   SARSTEDT
Microtubes with thread in special sterile packaging
Catalog # SARS72.609.711
Supplier:  SARSTEDT
Microtubes with thread in special sterile packaging
Microcentrifuge tubes with skirted base, sterile, 2,0 ml, 500/double bag, 2500/case
Price
149,00 €
Log in for availability
1 * 2.500 items
Product Details & Documents
PP, translucent
  • With or without skirted base
Each bag is sub-packaged into a second additional bag. Removing the outer bag leaves a complete and sterile bag packaging for convenient processing in clean rooms or sterile environments. This packaging option is ideal for users working under clean room conditions to fill precious reagents into tubes.
Specifications
  • Packed:
    500/double bag, 2500/case
  • Capacity:
    2,0 ml
  • Description:
    Microcentrifuge tubes with skirted base, sterile
Accessories
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.720
    Micro tubes
    Yellow
    Log in for availability
    1 * 5.000 items - 341,00 €
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.721
    Micro tubes
    Red
    Log in for availability
    1 * 5.000 items - 341,00 €
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.723
    Micro tubes
    Blue
    Log in for availability
    1 * 5.000 items - 341,00 €
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.725
    Micro tubes
    Neutral
    Log in for availability
    1 * 5.000 items - 358,00 €
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.726
    Micro tubes
    Green
    Log in for availability
    1 * 5.000 items - 341,00 €
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.727
    Micro tubes
    Orange
    Log in for availability
    1 * 5.000 items - 288,00 €
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.729
    Micro tubes
    Brown
    Log in for availability
    1 * 5.000 items - 288,00 €
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.728
    Micro tubes
    Black
    Log in for availability
    1 * 5.000 items - 288,00 €
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.757
    Micro tubes
    White
    Log in for availability
    1 * 5.000 items - 288,00 €
Product Information
  • Catalog #
    For
    Colour
    Availability
    Price
  • Colour coded screw caps, PP, sterile
    Colour coded screw caps, PP, sterile
    SARS65.716.755
    Micro tubes
    Violet
    Log in for availability
    1 * 5.000 items - 288,00 €