Orders LinkContactUsLinkComponent
Screw caps for micro tubes
Screw caps for micro tubes
Screw caps for micro tubes, Black
Catalog # SARS65.716.007
Supplier:   SARSTEDT
Screw caps for micro tubes
Catalog # SARS65.716.007
Supplier:  SARSTEDT
Screw caps for micro tubes
Screw caps for micro tubes, Black
Price
51,70 €
Log in for availability
1 * 1.000 items
Product Details & Documents
PP, coloured.
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw cap with O-ring seal.
Specifications
  • Colour:
    Black
  • Description:
    Screw caps for micro tubes
  • Seq:
    02