Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Tubes
Microtubes
Accessories for micro tubes
Accessories for micro tubes
Print
Accessories for micro tubes
Colour coded inserts, violet
Catalog # SARS65.713.007
Supplier:
SARSTEDT
Accessories for micro tubes
Catalog # SARS65.713.007
Supplier:
SARSTEDT
Expand image
Return to Top
Accessories for micro tubes
Colour coded inserts, violet
Specifications
Add to cart
Jump to:
Specifications
Price
13,30 €
Log in
for availability
Unit of Measure
1 * 1.000 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Description:
Colour coded inserts, violet