Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Tubes
Microtubes
Accessories for micro tubes
Print
Accessories for micro tubes
Colour coded inserts, red
Catalog # SARS65.713.002
Supplier:
SARSTEDT
Accessories for micro tubes
Catalog # SARS65.713.002
Supplier:
SARSTEDT
Return to Top
Accessories for micro tubes
Colour coded inserts, red
Specifications
Jump to:
Specifications
Price
72,80 €
Log in
for availability
1 * 5.000 items
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Description:
Colour coded inserts, red