0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Tubes
Microtubes
Accessories for micro tubes
Accessories for micro tubes
Colour coded inserts, neutral
SARS65.713
:
SARSTEDT
Accessories for micro tubes
SARS65.713
:
SARSTEDT
Accessories for micro tubes
Colour coded inserts, neutral
66,50 €
1 * 5.000 items
-
+
Description:
Colour coded inserts, neutral