Wire basket, round
240×180 mm
Catalog # DELTR-250
Supplier: DELTALAB
Jump to:
Product Details & Documents
Stainless steel.
These round baskets feature a folding handle for more convenience. The base of these baskets is reinforced by a thick mesh for added strength.
Ordering information: Models with inner crossed dividers are available.
Documents
Certificates
Search by Lot NumberSpecifications
- Ø×H:240×180 mm
- Seq:04