Order Entry
Export
Orders LinkContactUsLinkComponent
Diphenylsilane ≥97%

Diphenylsilane ≥97%

Glass bottle for solids, 25 g
Catalog # A10884.14
Irritant
Diphenylsilane ≥97%
Catalog # A10884.14
CAS Number:  775-12-2
Diphenylsilane ≥97%
Diphenylsilane ≥97%

Glass bottle for solids, 25 g

CAS Number:  775-12-2
Price
157,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 775-12-2 Molecular Formula: C12H12Si Formula Weight: 184.32 Flash Point: 98°(208°F) Refractive Index: 1.5790 Boiling Point: 95-97°/13mm Beilstein Reference: 2935887 MDL No.: MFCD00003002

  • CAS Number:775-12-2
  • Formula:C₁₂H₁₂Si
  • MDL Number:MFCD00003002
  • Molecular Weight:184.31 g/mol
  • Boiling Pt:95…97 °C (13 mmHg)
  • Density:0.996
  • Flash Pt:98 °C (208 °F)
  • Storage Temperature:Ambient
  • EINECS:212-271-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications