Order Entry
Export
Orders LinkContactUsLinkComponent
Serological pipettes

Serological pipettes

25 ml, 0,2 ml, +, Red, Individually wrapped, 200/case
Catalog # SARS86.1685.001
Supplier:   SARSTEDT
Serological pipettes
Catalog # SARS86.1685.001
Supplier:  SARSTEDT
Serological pipettes

25 ml, 0,2 ml, +, Red, Individually wrapped, 200/case

Price
108,00 €
Log in for availability
1 * 200 items
Product Details & Documents

PS, graduated, sterile.

  • Plugged
  • Easy identification by colour code
  • Larger pipetting volume due to negative graduation
  • Optimised mouth piece for universal fit in most common pipettors
  • Easy to open, static-free packaging
Specifications
  • Colour code:
    Red
  • Packed:
    Individually wrapped, 200/case
  • Sterile:
    +
  • Capacity:
    25 ml
  • Division:
    0,2 ml