Orders LinkContactUsLinkComponent
96-Well micro test plates
96-Well micro test plates
Micro test plate with flat bottom, with lid, sterile, Clear, 96, +, 1/bag
Catalog # SARS82.1581.001
Supplier:   SARSTEDT
96-Well micro test plates
Catalog # SARS82.1581.001
Supplier:  SARSTEDT
96-Well micro test plates
Micro test plate with flat bottom, with lid, sterile, Clear, 96, +, 1/bag
Price
93,30 €
Log in for availability
1 * 50 items
Product Details & Documents
Clear PS
  • Compatible with all common microplate dispensers, readers, and washers
  • Excellent and consistent optical quality
  • Optimal surface
  • Uniform design
  • Choice of flat, round or conical wells
Specifications
  • No. of wells:
    96
  • Packed:
    1/bag
  • Sterile:
    +
  • Colour:
    Clear
  • Description:
    Micro test plate with flat bottom, with lid, sterile
Accessories
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Lids for micro test plates, PS
    Lids for micro test plates, PS
    SARS82.1584
    Clear
    Log in for availability
    1 * 100 items - 43,60 €