Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Closures
Test/Sample Tube Closures
Screw caps for test tubes
Screw caps for test tubes
Print
Screw caps for test tubes
with gasket, 12 mm O.D., Red
Catalog # 212-0646
Supplier:
Greiner Bio-One
Screw caps for test tubes
Catalog # 212-0646
Supplier:
Greiner Bio-One
Expand image
Return to Top
Screw caps for test tubes
with gasket, 12 mm O.D., Red
Specifications
Add to cart
Jump to:
Specifications
Price
48,60 €
Log in
for availability
Unit of Measure
1 * 500 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
For tubes:
12 mm O.D.
Colour:
Red
Description:
with gasket
Seq:
12