1,2-Dibromoethane ≥99%
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 106-93-4 Molecular Formula: C2H4Br2 Formula Weight: 187.87 Flash Point: None Refractive Index: 1.5385 Melting Point: 9-10° Boiling Point: 132° Merck Reference: 14,3796 UN No.: 1605 MDL No.: MFCD00000233
- CAS Number:106-93-4
- Formula:BrH₂CCH₂Br
- MDL Number:MFCD00000233
- Molecular Weight:187.86 g/mol
- Merck Index:14,3796
- Boiling Pt:132 °C
- Melting Pt:9…10 °C
- Density:2.179
- Storage Temperature:Ambient
- EINECS:203-444-5
- UN:1605
- ADR:6.1,I
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g



