Orders LinkContactUsLinkComponent
Lids for Nunc™ microplates
Lids for Nunc™ microplates
Lid, PS, with cut-off corners and evaporation barrier, for use with 384-well plates, Clear, 7,9 mm, Sterile
Catalog # NUNC264616
   Low Manufacturing Impact
Lids for Nunc™ microplates
Catalog # NUNC264616
   Low Manufacturing Impact
Lids for Nunc™ microplates
Lids for Nunc™ microplates
Lid, PS, with cut-off corners and evaporation barrier, for use with 384-well plates, Clear, 7,9 mm, Sterile
Price
460,00 €
Log in for availability
1 * 180 items
Product Details & Documents
Specifications
  • Sterility:
    Sterile
  • Colour:
    Clear
  • Description:
    Lid, PS, with cut-off corners and evaporation barrier, for use with 384-well plates
  • Height:
    7,9 mm
  • Seq:
    09
  • Cat. No.:
    NUNC264616
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact