Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER PFRG FOR L-8500, L-8800, L-8900 1 * 1 L
Catalog # B29001
Supplier:
VWR Chemicals
Expand image
BUFFER PFRG FOR L-8500, L-8800, L-8900 1 * 1 L
Catalog # B29001
Supplier:
VWR Chemicals