Order Entry
Export
Orders LinkContactUsLinkComponent
Vacuum aspiration systems

Vacuum aspiration systems

Vacuum aspiration system, with base plate, holder and 4 l bottle
Catalog # 512-3360
Vacuum aspiration systems
Catalog # 512-3360
Vacuum aspiration systems
Vacuum aspiration systems

Vacuum aspiration system, with base plate, holder and 4 l bottle

Price
1 470,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Vacuum aspiration system. A plastic supporting plate with carrying handles and attachments for holding the vacuum-proof suction flask make the system compact and mobile at any time.

Specifications
  • Description:
    Vacuum aspiration system, with base plate, holder and 4 l bottle
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Foot switch with a 230 V socket
    Foot switch with a 230 V socket
    442-9468
    Log in for availability
    1 * 1 items - 371,00 €