DNA ladders, SimplyLoad™
SimplyLoad™ 100 bp DNA ladder: 32 ng/µl, 10 fragments from 100 bp to 1,000 bp, in 100 bp increments, 100 Assays
Catalog # LONZ50327
Supplier: LONZA
Jump to:
Product Details & Documents
Lonza SimplyLoad™ DNA ladders are ready to load directly onto the gel to maximise time and convenience. Each band has an ideal 50% G-C content for rapid, precise sizing and sharp resolution. Free from contaminating plasmid DNA so they have minimal background.
- Ready to load
- Storage temperature: 2 to 8 °C
Specifications
Specifications
- Pk:100 Assays
- Description:SimplyLoad™ 100 bp DNA ladder: 32 ng/µl, 10 fragments from 100 bp to 1,000 bp, in 100 bp increments



