Orders LinkContactUsLinkComponent
Screw caps for tubes
Screw caps for tubes
PP, 13-415
Catalog # DELT617100
Supplier:   DELTALAB
Screw caps for tubes
Catalog # DELT617100
Supplier:  DELTALAB
Screw caps for tubes
PP, 13-415
Price
361,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Screw caps made of polypropylene and aluminium with an internal liner made of rubber.
  • Both models of caps are fully autoclavable and are pharmaceutical quality
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw caps for round bottom borosilicate glass tubes and flat bottom soda glass tubes.
Documents
Specifications
  • Thread:
    13-415
  • Material:
    PP
  • Seq:
    02