Order Entry
Export
Orders LinkContactUsLinkComponent
Adhesion slides, HistoBond®+

Adhesion slides, HistoBond®+

Adhesion slides with frosted ends and marking side (20 mm), Pink
Catalog # MARI0810441
Supplier:   MARIENFELD
Adhesion slides, HistoBond®+
Catalog # MARI0810441
Supplier:  MARIENFELD
Adhesion slides, HistoBond®+
Adhesion slides, HistoBond®+

Adhesion slides with frosted ends and marking side (20 mm), Pink

Price
29,85 €
Log in for availability
1 * 100 items
Product Details & Documents

These adhesion slides are composed of silane treated surfaces and 90° ground edges.

  • Coloured marking areas on one end
  • Specially used for applications of colour coding
  • Improved identification of specimens

Certifications: IVD directive 98/79/EC

Specifications
  • Colour:
    Pink
  • Description:
    Adhesion slides with frosted ends and marking side (20 mm)