Order Entry
Export
Orders LinkContactUsLinkComponent
pH buffers, ready to use

pH buffers, ready to use

Technical buffer STP-10 Trace, 10,01, Plastic bottle, 50 mL
Catalog # WTWA108722
Supplier:   WTW
pH buffers, ready to use
Catalog # WTWA108722
Supplier:  WTW
CAS Number:  
pH buffers, ready to use
pH buffers, ready to use

Technical buffer STP-10 Trace, 10,01, Plastic bottle, 50 mL

CAS Number:  
Price
17,10 €
Log in for availability
1 * 50 mL
Product Details & Documents

Every bottle has its own lot reference with the expiration date.

  • Shelf life up to 3 years (unopened)
  • Reference temperature at 25 °C
  • Economic consumption
  • Easiest handling by use of the dosing vessel
  • One litre container with a separate compartment for the sample

Accuracy is ±0,03 pH

Certifications: Traceable to NIST.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Description:
    Technical buffer STP-10 Trace
  • pH value:
    10,01

Specifications