RNase A
5 mL
Supplier: OMEGA BIO-TEK
Jump to:
Product Details & Documents
Ribonuclease A (RNase A) is endonuclease-free and quality controlled for use in plasmid purification procedures for digestion of RNA.
Documents
Specifications
Specifications
- Pk:5 mL
- Cat. No.:AC118
- Concentration:25 mg/ml
- Enzyme name:RNase A