Order Entry
Export
Orders LinkContactUsLinkComponent
Melamine 99%

Melamine 99%

Glass bottle for solids, 2.500 g
Catalog # A11295.0E
Melamine 99%
Catalog # A11295.0E
CAS Number:  108-78-1
Melamine 99%
Melamine 99%

Glass bottle for solids, 2.500 g

CAS Number:  108-78-1
Price
84,80 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 99% CAS No.: 108-78-1 Molecular Formula: C3H6N6 Formula Weight: 126.12 Melting Point: ca 350° dec. Beilstein Reference: 124341 Merck Reference: 14,5811 MDL No.: MFCD00006055

Synonyms:  2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine
  • CAS Number:108-78-1
  • Formula:C₃H₆N₆
  • MDL Number:MFCD00006055
  • Molecular Weight:126.12 g/mol
  • Merck Index:14,5811
  • Melting Pt:∼ 350 °C (dec.)
  • Density:1.573
  • Flash Pt:>280 °C
  • Storage Temperature:Ambient
  • EINECS:203-615-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications