Agarose, SeaKem® LE
Agarose, SeaKem® LE
Catalog # 733-1536
Supplier: LONZA
Jump to:
Product Details & Documents
SeaKem® LE is a high gel strength, low EEO, standard gelling temperature agarose perfect for electrophoresis and other molecular biology applications.
- Quality tested to certify performance
- High gel strength
- Low EEO
- Standard melting temperature
- DNase- and RNase-free
SeaKem® LE agarose is ideal for routine analysis of nucleic acids by gel electrophoresis and blotting. Each SeaKem LE gel sharply resolves DNA and provides consistent resolution from lot-to-lot. This molecular biology grade agarose has no detectable DNase or RNase activity and forms strong gels with low background upon ethidium bromide, SYBR® Green, or GelStar™ staining. SeaKem® LE is also recommended for protein gel electrophoresis in applications such as Ouchterlony and radial immunodiffusion (RID).
Specifications
- Description:Agarose, SeaKem® LE
- Seq:01