Order Entry
Export
Orders LinkContactUsLinkComponent
Drums with screw cap

Drums with screw cap

5 L, 44 mm, 240 mm
Catalog # DELT202605
Supplier:   DELTALAB
Drums with screw cap
Catalog # DELT202605
Supplier:  DELTALAB
Drums with screw cap

5 L, 44 mm, 240 mm

Price
10,40 €
Log in for availability
1 * 1 items
Product Details & Documents

Drums, made of HDPE, provided with screw cap with grip. Can be used for the transport of hazardous, toxic and noxious products.

  • Stackable, with security cap
  • Good chemical resistance
  • Four flat sides for sticking information labels

Documents

Specifications
  • Ø opening:
    44 mm
  • Capacity:
    5 L
  • Height:
    240 mm
  • Seq:
    01