Order Entry
Export
Orders LinkContactUsLinkComponent
 

Photographic trays

350×300×85 mm
  DELT19283
 :   DELTALAB
Photographic trays
  DELT19283
 :  DELTALAB
Photographic trays

350×300×85 mm

 
99,90 €
   
1 * 1 items
 

Photographic trays made of PVC.

  • Temperature resistance from −20 to +80 °C

With ribbed interior surface.

 
  • Seq:
    04
  • W×D×H:
    350×300×85 mm